missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST8SIA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584205
This item is not returnable.
View return policy
Description
Immunogen sequence: SVIQRAFEDL VNATWREKLL QRLHSLNGSI LWIPAFMARG GKERVEWVNE Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 58%.
The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29.
Specifications
| ST8SIA2 | |
| Polyclonal | |
| Unconjugated | |
| ST8SIA2 | |
| AI323367; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; cb394; HsT19690; id:ibd5161; MGC116854; MGC116857; N-glycan alpha 2,8-sialyltransferase; polysialic acid synthase; sialyltransferase 8; sialyltransferase 8 (alpha-2 8-sialytransferase) B; sialyltransferase 8 (alpha-2, 8-sialyltransferase) B; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; sialyltransferase 8B; Sialyltransferase St8Sia II; sialyltransferase X; sialytransferase St8Sia II; SIAT 8B; siat8; SIAT8B; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2; ST8Sia II; ST8SIA2; st8sia2 protein; ST8SiaII; ST8SIA-II; STX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8128 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92186 | |
| ST8SIA2 | |
| Recombinant protein corresponding to Human ST8SIA2. Recombinant protein control fragment (Product #RP-101326). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction