missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody is specific to ST8SIA2.
The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29.
Specifications
Specifications
| Antigen | ST8SIA2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ST8SIA2 |
| Gene Accession No. | O35696, Q07977, Q92186 |
| Gene Alias | Alpha 2,8 sialyltransferase 8B, HsT19690, Sialyltransferase 8B, Sialyltransferase X, Sialytransferase St8Sia II, SIAT8 B, SIAT8B, ST8SIA II, ST8SIA2, ST8SiaII, STX |
| Gene Symbols | ST8SIA2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?