missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST8SIA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112727
This item is not returnable.
View return policy
Description
ST8SIA5 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| ST8SIA5 | |
| Polyclonal | |
| Unconjugated | |
| ST8SIA5 | |
| Alpha-2,8-sialyltransferase 8E; Sialyltransferase 8E; sialyltransferase 8E (alpha-2, 8-polysialytransferase); sialyltransferase St8Sia V; SIAT8E; SIAT8-E; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 5; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5; ST8SIA5; ST8SiaV | |
| Rabbit | |
| Protein G | |
| RUO | |
| 29906 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O15466 | |
| ST8SIA5 | |
| Recombinant Human Alpha-2,8-sialyltransferase 8E protein (101-376AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction