missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Stabilin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550834
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Stabilin 1 protein.
This gene encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 fasciclin, 16 epidermal growth factor (EGF)-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluronan-binding Link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node.
Specifications
| Stabilin 1 | |
| Polyclonal | |
| Unconjugated | |
| STAB1 | |
| CLEVER-1; common lymphatic endothelial and vascular endothelial receptor-1; fasciclin egf-like, laminin-type egf-like, and link domain-containing scavenger receptor-1; fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 1; FEEL1; FEEL-1; FEEL-1MS-1 antigen; FELE-1; FEX1; KIAA0246; MFEEL-1; mKIAA0246; MS-1; MS-1 antigen; SCARH2; scavenger receptor FEEL-1; STAB1; STAB-1; stabilin 1; stabilin-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23166 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NY15 | |
| STAB1 | |
| Synthetic peptide corresponding to residues near the C terminal of human stabilin 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction