missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stabilin-2 Antibody (OTI2E12), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74373B
This item is not returnable.
View return policy
Description
Stabilin-2 Monoclonal specifically detects Stabilin-2 in Human samples. It is validated for Western Blot.
Specifications
| Stabilin-2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| STAB2 | |
| Recombinant protein fragment corresponding to amino acids 1914-2194 of human Stabilin-2 (NP_060034) produced in E.coli. | |
| 0.1 mL | |
| 55576.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2E12 | |
| Western Blot | |
| CD44-like precursor FELL, DKFZP434E0321, FAS1 EGF-like and X-link domain containing adhesion molecule-2, FAS1 EGF-like and X-link domain-containing adhesion molecule 2, fasciclin egf-like, laminin-type egf-like, and link domain-containing scavengerreceptor-2, Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavengerreceptor 2, FEEL2, FEEL-2, FELE-2, FELL, FEX2, HAREFELL2, hepatic hyaluronan clearance receptor, Hyaluronan receptor for endocytosis, hyaluronic acid receptor for endocytosis, STAB-2, stabilin 2, stabilin-2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction