missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Stablcal™ Stabilized Formazine Turbidity Standard 0.1 NTU

Product Code. 11996738
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11996738 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11996738 Supplier Hach Lange™ Supplier No. 2659749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hach Lange Stablcal Stabilized Formazine Turbidity Standard 0.1 NTU, Volume: 100mL, Stablcal Stabilized Formazine Turbidity Standard <0.1 NTU

Hach Lange Stablcal Stabilized Formazine Turbidity Standard 0.1 NTU, Volume: 100mL, Stablcal Stabilized Formazine Turbidity Standard <0.1 NTU

TRUSTED_SUSTAINABILITY

Specifications

Type Stablcal Stabilized Formazine Turbidity Standard
Turbidity <0.1 NTU
Volume (Metric) 500 mL
compliance-icons
Product Identifier
  • StablCal Formazin Turbidity Standard
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: methenamine, sodiumsulphate, formaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.