missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAC Monoclonal Antibody (2C5)
GREENER_CHOICE

Product Code. 15508862
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15508862 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508862 Supplier Invitrogen™ Supplier No. MA520035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: LAQRTKKGSS GSGSDSPHRT STSDLVEVPE EANGPGGGYD LRKRSNSVFT YPENGTDDFR DPAKNINHQG SLSKDPLQMN TYVALYKFVP QENEDLEMRP GDIITLLEDS.

Promotes expression of the ion channel CACNA1H at the cell membrane, and thereby contributes to the regulation of channel activity. Plays a minor and redundant role in promoting the expression of calcium channel CACNA1S at the cell membrane, and thereby contributes to increased channel activity. Slows down the inactivation rate of the calcium channel CACNA1C. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene STAC
Gene Accession No. Q99469
Gene Alias SH3 and cysteine rich domain; SH3 and cysteine-rich domain-containing protein; Src homology 3 and cysteine-rich domain-containing protein; src homology three (SH3) and cysteine rich domain; Stac; STAC1
Gene Symbols STAC
Host Species Mouse
Immunogen STAC (AAH20221, 209 a.a. ~ 318 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6769
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.