Learn More
Vitlab™ Stackable Graduated Beaker
Description
- Highly transparent graduated beakers
- Volume is equally visible for left- and right-handers
- To preserve markings, cleaning at no higher than 60°C is recommended
Specifications
Specifications
| Color | Transparent |
| Diameter (Metric) Outer | 100 mm |
| Capacity (Metric) | 500 mL |
| Material | Polypropylene |
| Height (Metric) | 140 mm |
| Quantity | 1 Ea. |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Graduated | Yes |
| Scale | Black Printed Scale |