Learn More
Thermo Scientific™ Stained Proteus Suspensions
Rapidly detect and quantify reckettsial antibodies in sera with this suspension.
Brand:  Thermo Scientific™ R30953801
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15248674
  • € 95.71 / 5mL
Estimated Shipment
28-07-2020
Add to basket
Description
Description
Use Stained Proteus Suspensions for the quantitative detection of rickettsial antibodies in sera for epidemiological and diagnostic purposes in the investigation of rickettsial infections (Weil-Felix test)1, 2.
  • Simple, easy to use tests
  • Provide accurate, reliable results in as little as one minute
The tests are based on the fact that somatic constituents of some strains of Proteus are shared with some species of Rickettsia. Sera from people with some rickettsial infections therefore will agglutinate suspensions of the Proteus strains.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
Proteus OXK
Stained Proteus Suspensions
Stained Proteus Suspensions
2° - 8°C