Learn More
Grant Instruments™ Stainles Steel Clamp for Flasks
Click to view available options
Holds:
100 mL flask
1000 mL flask
250 mL flask
50 mL flask
500 mL flask
Unit Size:
Each
Description
Clamp, stainless steel,flask, UP-168, platform,ES-20/80, Grant Instruments
Specifications
Specifications
| Holds | 250 mL flask |
| Product Type | Clamp |
| Description | Tight fit clamp 250 mL - 85 mm dia. for UP-168 |
| For Use With (Equipment) | UP-168 |