Learn More
Caframo™ 3 Blade Turbines
GREENER_CHOICE
Product Code. 12947126
Click to view available options
Diameter (Metric) Shaft:
8 mm
9.5 mm
Unit Size:
Each
Product Code. 12947126
Brand: Caframo™ A553
Only null left
Add to basket
Add to basket
This item is not returnable. Afficher la politique du retour.
Stainless steel 3 blade turbines in multiple sizes for use with Caframo™ and Fisherbrand™ Overhead Stirrers. Ideal for high shear mixing with moderate vortexing
  • Electropolished 316L stainless steel
  • Chemically inert and corrosion resistant
  • High shear with moderate vortexing characteristics
  • Set screws included
Spécification
Diameter (Metric) 50 mm
Diameter (Metric) Shaft 9.5 mm
Type Radial Flow Turbine
Material Stainless Steel (316L)