Learn More
Schneider Gerd™ Stainless Steel 4301 Sharp Forceps, Curved
Product Code. 11387364
Click to view available options
Length (Metric):
130 mm
Unit Size:
Each
Product Code. 11387364
Brand: Schneider Gerd™ 142
Only null left
Add to basket
Add to basket
Schneider Gerd™ Stainless Steel 4301 Sharp Forceps, Curved is used in dissecting work.
  • Made of Stainless steel 4301
Specifications
Length (English) 5 in.
Material Stainless Steel (4301)
Length (Metric) 130 mm
Product Type Forceps, Sharp
Style Curved