Learn More
Schneider Gerd™ Stainless Steel 4301 Sharp Forceps, Straight
Click to view available options
Length (Metric):
130 mm
Unit Size:
Each
Description
- Made of Stainless steel 4301
Specifications
Specifications
| Length (English) | 5 in. |
| Material | Stainless Steel (4301) |
| Length (Metric) | 130 mm |
| Product Type | Forceps, Sharp |
| Style | Straight |