Learn More
Schneider Gerd™ Stainless Steel 4301 Thumb Forceps, Straight
Click to view available options
Length (Metric):
200 mm
Unit Size:
Each
Description
- Made of Stainless steel 4301
Specifications
Specifications
| Length (English) | 7.8 in. |
| Material | Stainless Steel (4301) |
| Length (Metric) | 200 mm |
| Product Type | Forceps, Thumb |
| Style | Straight |