Learn More
Bochem™ Stainless Steel Beakers
Product Code. 10729621
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Includes:
Rim
Rim and Spout
Rim, Handle
Rim, Spout
Rim, Spout, Handle
Unit Size:
Each
Product Code. 10729621
Brand: Bochem™ 8506
Only null left
Add to basket
Add to basket
Highly resistant and durable stainless steel beaker. Bochem™ Stainless Steel Beakers include rims, spouts, and handles in various model combinations.
  • 18/10 stainless steel construction.
  • Resistant and durable.
  • Contact bottom for heating plates and magnetic stirrers.
Specifications
Capacity (Metric) 5000 mL
Material Stainless Steel (18/10)
Includes Rim
Type Beaker