Learn More
VACUUBRAND™ Stainless Steel Bellow Sealed Angle Valve VE
Click to view available options
Size (Metric):
16 mm dia.
25 mm dia.
40 mm dia.
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel |
| Quantity | 1 Each |
| Size (Metric) | 16 mm dia. |
| Type | Bellow Sealed Angle Valve VE |