Learn More
B Braun™ Bent Tip Micro Tweezers with Round Handle
Product Code. 10539033
Change view
Click to view available options
Length (Metric):
120 mm
150 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10539033 120 mm Each
10505824 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10539033 Supplier B Braun™ Supplier No. FD281R
  • ONLINE EXCLUSIVE PRICE
    € 324.00
    € 312.90 / Pack of 1
    Save € 11.10
    3% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
missing translation for 'validMainframeMsg'
Use B Braun™ Bent Tip Micro Tweezers with Round Handle as a versatile tool for patient needs that dictate their use.
  • Stainless Steel
  • Bent Tip
  • 1 per pack
  • Round handle
Specifications
Product Type Tweezers, Micro
Material Stainless Steel
Quantity 1 Pack
Tip Style Bent
Length (Metric) 120 mm
Width (Metric) 0.4 mm