Learn More
Bochem™ Stainless Steel Blunt Tip Forceps
Product Code. 10177430
Click to view available options
Length (Metric):
120 mm
Unit Size:
Each
Product Code. 10177430
Brand: Bochem™ 1641
Only null left
Add to basket
Add to basket
Type 2. Bochem™ Stainless Steel Blunt Tip Forceps can be used for precision applications.
  • Points without ridges
  • Selftensing
Specifications
Tip Style Curved
No. per Pack 1 Pack
Material Stainless Steel
Length (Metric) 120 mm
Product Type Forceps, Precision
Type Forceps