Learn More
Memmert™ Stainless Steel Bottom Drip Tray for IN450plus, 15 mm
Product Code. 17577745 Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 17577745
Brand: Memmert™ IN450plusB04362
Only null left
Add to basket
Add to basket
Non-perforated bottom drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) IN450plus Incubator
Material Stainless Steel
Type Bottom Drip Tray
Height (Metric) Rim: 15 mm