Learn More
Memmert™ Stainless Steel Bottom Drip Tray for IN750plus, Reinforced Chamber Option
Product Code. 17557935 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN750plus Incubator
Unit Size:
Each
Product Code. 17557935
Brand: Memmert™ IN750plusB34055
Only null left
Add to basket
Add to basket
Non-perforated bottom drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) IN750plus Incubator
Material Stainless Steel
Type Bottom Drip Tray
Height (Metric) Rim: 15 mm