Learn More
Sampling Systems™ Stainless Steel Buckets with Pouring Spouts
Product Code. 15839651 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
10 L
15 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15839651 15 L Each
15829651 10 L Each
15849651 - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15839651 Supplier Sampling Systems™ Supplier No. A40415
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These high quality buckets are made from heavy gauge 316L stainless steel.
  • Fitted with a skirt band to make buckets as stable as possible
  • Crevice-free interior
  • Not graduated
  • Swing handle
  • Lids available as an accessory
Specifications
Product Type Bucket with Pouring Spout
Capacity (Metric) 15 L
Graduations No
Material Stainless Steel (316L)
Includes Swing Handle, Bottom Skirt