Learn More
Sampling Systems™ Stainless Steel Buckets with Pouring Spouts
Product Code. 15849651 Shop All Sampling Systems Products
Click to view available options
Unit Size:
Each
Capacity (Metric):
10 L
15 L
Product Code. 15849651
Brand: Sampling Systems™ A40420
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These high quality buckets are made from heavy gauge 316L stainless steel.
  • Fitted with a skirt band to make buckets as stable as possible
  • Crevice-free interior
  • Not graduated
  • Swing handle
  • Lids available as an accessory
Specifications
Product Type Bucket with Pouring Spout
Graduations No
Material Stainless Steel (316L)
Includes Swing Handle, Bottom Skirt