Learn More
Corning™ Stainless Steel Digital Dry Bath Heater
Product Code. 11912021 Shop All Corning Products
Click to view available options
Amperage:
1 A
Model:
Dual Block
Unit Size:
Each
Product Code. 11912021
Brand: Corning™ 6786DB
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Note: Blocks are sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Controller Type Microprocessor
Material Stainless Steel
Temperature Range (Metric) 5°C to 150°C
Display LED
Model Dual Block
No. of Blocks 2
Temperature Accuracy ±0.3°C
Temperature Stability 0.1°C
Temperature Uniformity ±0.2°C
Dimensions (L x W x H) 20 x 26.5 x 8.3 cm
Length (Metric) 20 cm
Width (Metric) 26.5 cm
Height (Metric) 8.3 cm
Weight (Metric) 2.2 kg
Electrical Requirements 230 V, 50/60 Hz
Voltage 230 V
Amperage 1 A
Frequency 50/60 Hz