Learn More
RETSCH Stainless Steel Ring Sieve For Ultra Centrifugal Mill ZM 200
Shop All Retsch Products
Click to view available options
Pore Size:
2 mm
10 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 10 mm |
| Type | Ring Sieve |
| Material | Stainless Steel |
| For Use With (Equipment) | Ultra Centrifugal Mill ZM 200 |