Learn More
Schneider Gerd™ Stainless Steel Double Micro-spatula
Product Code. 10060971
Click to view available options
Length (Metric):
100 mm
130 mm
150 mm
210 mm
Width (Metric):
2 mm
3 mm
4 mm
5 mm
6 mm
Unit Size:
Each
Product Code. 10060971
Brand: Schneider Gerd™ 3851
Only null left
Add to basket
Add to basket
Use this micro spatula for a variety of applications. Schneider Gerd™ Stainless Steel Double Micro-spatulas are ideal for use with a wide range of compounds.
  • Spatulas are ideal for working with powders, stirring and mixing
  • Perfect for working with miniscule amounts of chemicals
  • Two different end shapes, offer increased versatility
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Quantity 1 Ea.
Shape Double
Product Type Double Ended Micro Spatula
Width (Metric) 5 mm
Material Stainless Steel
Length (Metric) 130 mm