Learn More
Schneider Gerd™ Stainless Steel Double Micro-spatula
Click to view available options
Length (Metric):
100 mm
130 mm
150 mm
210 mm
Width (Metric):
2 mm
3 mm
4 mm
5 mm
6 mm
Unit Size:
Each
Description
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for working with miniscule amounts of chemicals
- Two different end shapes, offer increased versatility
- Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
| Quantity | 1 Ea. |
| Shape | Double |
| Product Type | Double Ended Micro Spatula |
| Width (Metric) | 2 mm |
| Material | Stainless Steel |
| Length (Metric) | 100 mm |