Learn More
Sampling Systems™ Stainless Steel Economy Scoops
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
120 mL
450 mL
1000 mL
1500 mL
2000 mL
Unit Size:
Each
Description
- Manufactured from spot welded 304 stainless steel
- With an attractive high polish
- Light duty construction
- Not suitable for GMP applications as there are crevices
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Product Type | Economy Scoop |
| Finish | High polish |
| For Use With (Application) | General industrial use |
| Material | Stainless Steel (AISI 304) |
| Length (Metric) | 315 mm |