Learn More
Sampling Systems™ Stainless Steel Economy Scoops
Product Code. 15889611 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
120 mL
450 mL
1000 mL
1500 mL
2000 mL
Unit Size:
Each
Product Code. 15889611
Brand: Sampling Systems™ A304450
Only null left
Add to basket
Add to basket
These Economy Scoops are ideal for general industrial use.
  • Manufactured from spot welded 304 stainless steel
  • With an attractive high polish
  • Light duty construction
  • Not suitable for GMP applications as there are crevices
Specifications
Capacity (Metric) 450 mL
Product Type Economy Scoop
Finish High polish
For Use With (Application) General industrial use
Material Stainless Steel (AISI 304)
Length (Metric) 255 mm