Learn More
RETSCH Stainless Steel Feed Hoppers
Product Code. p-8004947 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
For Chutes 15 and 40mm
Capacity (Metric):
2.8 L
400 L
Unit Size:
Each
Product Code. 10248491
Brand: RETSCH 03.785.0146
Only null left
Specifications
Product Type Feed Hopper
Capacity (Metric) 2.8 L
For Use With (Equipment) For Chutes 15 and 40mm
Material Stainless Steel