Learn More
Agencinox™ Stainless Steel Flat Bottom Dish
Click to view available options
Capacity (Metric):
20 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 5
Description
- Flat bottom stainless steel lotion bowls
- Autoclavable
- Available in a variety of sizes
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 20 mL |
| Material | Stainless Steel |
| Shape | Round |
| Type | Dish |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 40 mm |