Learn More
Usbeck Laborgeraete™ Stainless Steel Forceps
Product Code. p-8006520
Click to view available options
Length (Metric):
105 mm
115 mm
130 mm
160 mm
200 mm
Design:
Curved
Straight
Unit Size:
Each
Product Code. 10543952
Brand: Usbeck Laborgeraete™ 3030
Only null left
Specifications
Tip Style Fine
No. per Pack 1 Pack
Design Straight
Material Stainless Steel
Length (Metric) 105 mm
Dimensions (L x W) 105 L mm
Type Forceps