Learn More
Tenak™ Stainless Steel Freezer Rack, 130 mm Box Height
Shop All TENAK Products
Click to view available options
Positions:
6
8
10
12
15
Unit Size:
Each
Description
- For use with freezers
- Constructed of stainless steel
Specifications
Specifications
| Holds | Boxes |
| Material | Stainless Steel |
| Positions | 12 |
| For Use With (Equipment) | Freezers |
| Height (Metric) | 130 mm |
| Array | 3 x 4 |