Learn More
Memmert™ Stainless Steel Gable Cover, For One22
Highly durable and easy to maintain stainless steel gable cover. Memmert™ Gable Cover for Shaking Device/Peltier Cooling Unit fits shaking device (14/22 und 29/45) or Peltier cooling unit.
Brand: Memmert™ ONE22B02643
Additional Details : Weight : 0.01100kg
Description
- Stainless steel construction
- Durable
- Corrosion resistant
- Easy to clean
- Perfectly fitted
Specifications
14/22 and 29/45 Shaking Device, CDP 115 Peltier Cooling Unit Models 14 and 22 | |
Stainless Steel | |
3 Years |