Learn More
Memmert™ Stainless Steel Gable Cover, For One7
Highly durable and easy to maintain stainless steel gable cover. Memmert™ Gable Cover for Shaking Device/Peltier Cooling Unit fits shaking device (14/22 und 29/45) or Peltier cooling unit.
Brand:  Memmert™ ONE7B02641
Additional Details : Weight : 0.01100kg
Product Code. 16324057
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Stainless steel construction
  • Durable
  • Corrosion resistant
  • Easy to clean
  • Perfectly fitted
Specifications
Specifications
14/22 and 29/45 Shaking Device, CDP 115 Peltier Cooling Unit Model 7
Stainless Steel
3 Years
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers