Learn More
RETSCH Stainless Steel Grinding Balls
Shop All Retsch Products
Click to view available options
For Use With (Equipment):
MM 200 and MM 301
Planetary Ball Mill PM 400
Reaction Vials
Diameter (Metric):
3 mm
4 mm
5 mm
7 mm
15 mm
40 mm
Unit Size:
Each
Pack of 200
Description
- For use with MM 200 and MM 400 Mixer Mills
Specifications
Specifications
| Product Type | Grinding Balls |
| Diameter (Metric) | 7 mm |
| For Use With (Equipment) | MM 200 and MM 301 |
| Material | Stainless Steel |