Learn More
RETSCH Stainless Steel Grinding Jar
Product Code. 10099031 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
For CryoMill
For EMAX
For MM 200
For MM 301 Mixer mill
Capacity (Metric):
1.5 mL
5 mL
10 mL
25 mL
35 mL
50 mL
125 mL
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Capacity (Metric) Unit Size
10099031 For MM 301 Mixer mill 1.5 mL Each
10504041 For MM 200 1.5 mL Each
10034170 For MM 200 10 mL Each
10334172 For MM 301 Mixer mill 10 mL Each
15300974 For EMAX 125 mL Each
15370994 For CryoMill 35 mL Each
15370964 For EMAX 50 mL Each
10232672 For MM 301 Mixer mill 35 mL Each
10626275 For CryoMill 25 mL Each
10399770 For MM 200 5 mL Each
10160572 For MM 301 Mixer mill 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10099031 Supplier RETSCH Supplier No. 01.462.0230
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use these grinding jars for dry, wet, and cryogenic grinding of small samples. Retsch™ Stainless Steel Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Product Type Grinding Jar
Capacity (Metric) 1.5 mL
For Use With (Equipment) For MM 301 Mixer mill
Material Stainless Steel