Learn More
Alpaco™ Stainless Steel Clamps with Vertical Screws
Product Code. 11949667
Click to view available options
Size (Metric) Grip:
12 to 17 mm
17 to 22 mm
Unit Size:
Pack of 10
Product Code. 11949667
Brand: Alpaco™ INH 17/101618
Only null left
Add to basket
Add to basket
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Stainless Steel Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
  • Reusable
  • Resists pressure
  • No lubricant required for assembly
  • Smooth clamping to avoid tubing damage
Specifications
Quantity 10/Pk.
Type Clamp
Material Stainless Steel
Size (Metric) Grip 12 to 17 mm