Learn More
Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
Brand:  Bandelin Electronic™ 354
Additional Details : Weight : 25.00000kg
 View more versions of this product
Product Code. 10049360
  • € 222.00 / Each
Estimated Shipment: 26-04-2023
to see stock.
Add to basket
Description
Description
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
  • Available for a wide range of tank capacities
  • Available in a range of mesh sizes
  • Excellent for the cleaning of technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
Basket Insert
50 mm
Stainless Steel
275 x 245 x 50 mm
Ultrasonic Bath, RK 514/H
275 mm
245 mm
K14