Learn More
Schneider Gerd™ Stainless Steel Laboratory Forceps
Click to view available options
Length (Metric):
105 mm
130 mm
145 mm
300 mm
Tip Style:
Blunt
Fine
Unit Size:
Each
Description
- Stainless Steel Laboratory Forceps
- Ideal for dissection and general lab applications
Specifications
Specifications
| Tip Style | Blunt |
| Material | Stainless Steel |
| Length (Metric) | 300 mm |
| Product Type | Forceps |
| Quantity (No. per Case) | Each |
| Type | Forceps, Laboratory |