Learn More
Grant Instruments™ Stainless Steel Lid with Ring Sets
Click to view available options
For Use With (Equipment):
JBA12, JBN12, SAP12
JBA18, JBN18, SAP18
JBA5, JBN5, SAP5
SAP34
Unit Size:
Each
Description
- Stainless steel construction
- Flat lids contain 2, 4 or 6 ring sets.
- Variable hole diameter
- Accommodates taller vessels
Specifications
Specifications
| Description | 4-Ring Sets |
| For Use With (Equipment) | JBA12, JBN12, SAP12 |
| Material | Stainless Steel |