Learn More
Hamilton™ Custom Kel-F Hub Needles
Product Code. 11513274
Click to view available options
Needle Gauge:
10 G
14 G
16 G
17 G
20 G
22 G
22s G
25 G
28 G
30 G
Needle Point Style:
2, 3, or 4
5 or AS
Custom
Unit Size:
Pack of 6
Product Code. 11513274
Brand: Hamilton™ 775218
Design controlled by a series of universal standards which guarantees compatibility between manufacturers.
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style
  • Autoclavable
  • Gas sterilizable
  • For Kel-F hubs
  • Custom length
  • Stainless steel Luer tip
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge 22s G
Diameter (Metric) Inner 0.17 mm
Diameter (Metric) Outer 0.72 mm
Product Type Luer Lock Kel-F Hub Needle
Quantity 6/Pk.
Needle Point Style 5 or AS
For Use With (Equipment) LT and TLL Syringes and Connectors
Connection Type Standard Tapered