Learn More
VACUUBRAND™ Metal Vacuum Tubing With Small Flanges KF
Product Code. 10633762
Click to view available options
Tubing Size:
10
16
25
DN 40
Overall Length (Metric):
25 cm
250 mm
50 cm
75 cm
100 cm
Unit Size:
Each
Product Code. 10633762
Brand: VACUUBRAND™ 20673305
Only null left
Add to basket
Add to basket
A durable connection option for the laboratory that still provides flexibility.
  • Manufactured from stainless steel
  • Available in a range of sizes and lengths
TRUSTED_SUSTAINABILITY
Specifications
Description Single bending radius of 19
Diameter (Metric) Inner 10 mm
Material Metal
Diameter (Metric) Outer 10 mm
Overall Length (Metric) 250 mm
Packaging Quantity 1 Each
Tubing Size 10
Product Type Vacuum Tubing