Learn More
Bochem™ Stainless Steel Micro Clamp
Click to view available options
Type:
Micro Clamp
Unit Size:
Each
Specifications
Specifications
| Length (Metric) | 150 mm |
| Material | Stainless Steel |
| Type | Micro Clamp |
| Diameter (Metric) Rod | 15 mm |