Learn More
Cole-Parmer™ Stainless Steel Mixer Propellers
Product Code. 11720039 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (Metric):
Bore: 10 mm
Bore: 8 mm
Diameter (Metric) Fan:
51 mm
76 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Diameter (Metric) Fan unitSize
11720039 Bore: 10 mm 51 mm Each
11789929 Bore: 8 mm 51 mm Each
11820130 Bore: 8 mm 76 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11720039 Supplier Cole-Parmer™ Supplier No. 0455255
  • ONLINE EXCLUSIVE PRICE
    € 95.00
    € 86.41 / Pack of 1
    Save € 8.59
    9% Off
View all promotions
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
Use these three-blade propellers alone or mount several on a single shaft for thorough agitation. Stainless Steel Mixer Propellers are ideal for general-purpose mixing.
  • Propellers are made from electropolished 316 stainless steel
Specifications
Diameter (Metric) Bore: 10 mm
Diameter (Metric) Fan 51 mm
For Use With (Application) General-purpose mixing
Quantity 1 Each
Type Mixer Propeller