Learn More
B Braun™ Stainless Steel Syringe Replacement Needles
Product Code. 10320341
Click to view available options
Needle Gauge:
19 G
22 G
24 G
25 G
26 G
27 G
Length (Metric):
12 mm
16 mm
25 mm
30 mm
50 mm
Unit Size:
Pack of 100
Product Code. 10320341
Brand: B Braun™ 4665317
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replace needles on syringes with removable needles using Use B Braun™ Sterican™ Stainless Steel Syringe Replacement Needles. for relevant routine, pre- and post-op patient needs.
  • Sterican™
  • Blunt
  • Stainless steel
  • Latex-free
  • Sterile
Specifications
Material Stainless Steel
Needle Gauge 19 G
Diameter (Metric) Outer 1.1 mm
Latex Free Yes
Length (Metric) 30 mm
Product Type Needle
Sterility Sterile