Learn More
Sartorius™ Stainless Steel OIML Class F2 Reference Line Weight
Product Code. 10163981 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
50 kg
Case Material:
Plastic
Wood
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass Case Material unitSize
10163981 10 kg Wood Each
10123841 100 g Wood Each
10612022 10 g Plastic Each
10430272 1 g Plastic Each
10062211 1 kg Wood Each
10397631 200 g Wood Each
10748002 20 g Plastic Each
10451422 20 kg Wood Each
10255651 2 g Plastic Each
10103891 2 kg Wood Each
10397871 500 g Wood Each
10459691 50 g Wood Each
10614332 5 kg Wood Each
10163741 50 kg Wood Each
10634332 5 g Plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 10163981 Supplier Sartorius™ Supplier No. YCW71400
Stainless Steel OIML Class F2 Reference Line Weight without DAkkS Calibration Certificate.
  • Without DAkkS Calibration Certificate
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Case Material Wood
Mass 10 kg