Learn More
Nickel Electro™ Stainless Steel Perforated Tray
Shop All Nickel-Electro Products
Click to view available options
No. of Positions:
12
18
2
2 x 1000 mL, 6 x 500 mL, 15 x 100 mL, 35 x 25 mL
32
39
5
8
Product Type:
Perforated Tray
Universal Spring Tray
Unit Size:
Each
Description
- Supplied ready to assemble
- Stainless steel construction
- Suits glass/polypropylene Erlenmeyer flasks
- Tray simply drops onto shaker or rocking table platform and locates snugly
Specifications
Specifications
| Holds | 25 mL flask |
| Product Type | Perforated Tray |
| Material | Stainless Steel |
| Clamps | Yes |
| For Use With (Equipment) | NE5-28 Shaker Baths |
| No. of Positions | 39 |