Learn More
Sampling Systems™ Stainless Steel PharmaScoop™
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
10 mL
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1250 mL
1.7 L
2.5 L
5 L
Length (Metric):
75 mm
110 mm
180 mm
235 mm
240 mm
305 mm
320 mm
330 mm
340 mm
355 mm
Unit Size:
Each
Description
- Very high quality scoops
- Heavy duty construction
- Made from 316L Stainless Steel
- Crevice free
- Satin finish
- Rounded corners for easy cleaning
- GMP correct
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Product Type | Powder Scoop |
| Finish | Mirror Polish |
| Diameter (Metric) Outer | 50 mm |
| Material | Stainless Steel (316L) |
| Length (Metric) | 180 mm |