Learn More
Buerkle™ Stainless Steel PharmaSpoon™
Shop All Bürkle Products
Click to view available options
Length (Metric):
650 mm
Unit Size:
Each
Description
- For taking samples of powdery substances, intermediate products and material in the pharmaceutical industry
- Special materials with excellent surface qualities must be used for this to prevent cross-contamination
- Made of stainless steel AISI 316L (1.4404)
- Surface highly polished by hand (not electro-polished)
- Surface properties of the PharmaSpoon provide significant advantages
- Possible residues are even visible in the trace sector (μg sector)
- The surface is easy to clean
- The PharmaSpoon is also suitable for sterile sampling
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Spoon |
| Finish | High polish |
| For Use With (Application) | Foodstuffs |
| Material | Stainless Steel (V4A) |
| Length (Metric) | 650 mm |