Learn More
Stainless Steel powder, -140 mesh, Type 303-L, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 011084.18
Additional Details : Weight : 0.05000kg
Description
This offers moderate corrosion resistance. Alloy 303 is frequently used in applications that require parts to be heavily machined. It is used in nuts and bolts, aircraft fittings, gears and screws.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel powder, Type 303-L | |
MFCD00148499 | |
Insoluble in water. | |
Powder |
Fe:Cr:Ni; 70:17:13 wt% | |
50 g | |
−140 Mesh |
RUO – Research Use Only